Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCRL4
Entrez Gene ID
102170192
Gene Name
Fc receptor like 4
Gene Aliases
-
Chromosome Location
Chr.3: 107264513 - 107288860 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013976239.2 XP_013831693.2 6 - 7 1292 74

View Full Product Details