Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA11
Entrez Gene ID
102172209
Gene Name
serpin family A member 11
Gene Aliases
-
Chromosome Location
Chr.21: 58532368 - 58545397 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695351.3 XP_005695408.2 3 - 4 1157 75
XM_005695352.3 XP_005695409.2 3 - 4 1148 75
GenBank mRNA JO421733.2 - 2 - 3 506 75
JO434683.2 - 2 - 3 460 75

View Full Product Details