Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDRT1
Entrez Gene ID
102185393
Gene Name
CMT1A duplicated region transcript 1
Gene Aliases
FBXW10
Chromosome Location
Chr.19: 32673493 - 32704183 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013972619.2 XP_013828073.2 2 - 3 852 90

View Full Product Details