Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C4BPB
Entrez Gene ID
102178469
Gene Name
complement component 4 binding protein beta
Gene Aliases
-
Chromosome Location
Chr.16: 4281336 - 4294245 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690425.3 XP_005690482.1 2 - 3 548 90
GenBank mRNA JO421720.2 - 2 - 3 372 90
JO421721.2 - 2 - 3 372 90

View Full Product Details