Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAP4
Entrez Gene ID
102186756
Gene Name
A-kinase anchoring protein 4
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 16119650 - 16157123 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700732.3 XP_005700789.1 2 - 3 875 65
XM_013976472.2 XP_013831926.1 1 - 2 514 65
XM_013976477.2 XP_013831931.1 2 - 3 198 65
XM_013976478.2 XP_013831932.1 2 - 3 172 65
XM_018043992.1 XP_017899481.1 4 - 5 2068 65
XM_018043993.1 XP_017899482.1 4 - 5 2101 65
GenBank mRNA KA311530.1 - 2 - 3 80 65

View Full Product Details