Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQCF6
Entrez Gene ID
102189633
Gene Name
IQ motif containing F6
Gene Aliases
-
Chromosome Location
Chr.22: 49166020 - 49166445 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013973691.1 XP_013829145.1 1 - 1 321 72

View Full Product Details