Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IP6K3
Entrez Gene ID
102185047
Gene Name
inositol hexakisphosphate kinase 3
Gene Aliases
-
Chromosome Location
Chr.23: 40811955 - 40836106 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039298.1 XP_017894787.1 2 - 3 596 81
XM_018039299.1 XP_017894788.1 2 - 3 596 81

View Full Product Details