Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRPH2
Entrez Gene ID
102169531
Gene Name
peripherin 2
Gene Aliases
-
Chromosome Location
Chr.23: 32426997 - 32441228 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696314.1 XP_005696371.1 2 - 3 1108 76

View Full Product Details