Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT4
Entrez Gene ID
102170652
Gene Name
synaptotagmin 4
Gene Aliases
-
Chromosome Location
Chr.24: 12775729 - 12785834 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696966.3 XP_005697023.2 2 - 3 1097 101
GenBank mRNA JT728313.1 - 2 - 3 1024 101

View Full Product Details