Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCYOX1L
Entrez Gene ID
102172354
Gene Name
prenylcysteine oxidase 1 like
Gene Aliases
-
Chromosome Location
Chr.7: 49649982 - 49662586 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050178.1 XP_017905667.1 2 - 3 353 88
GenBank mRNA JO608948.2 - 4 - 5 1417 88

View Full Product Details