Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL21A1
Entrez Gene ID
102182838
Gene Name
collagen type XXI alpha 1 chain
Gene Aliases
-
Chromosome Location
Chr.23: 44384925 - 44639374 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039271.1 XP_017894760.1 12 - 13 1766 58
XM_018039272.1 XP_017894761.1 4 - 5 202 58

View Full Product Details