Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL21A1
Entrez Gene ID
102182838
Gene Name
collagen type XXI alpha 1 chain
Gene Aliases
-
Chromosome Location
Chr.23: 44384925 - 44639374 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039271.1 XP_017894760.1 13 - 14 1815 85
XM_018039272.1 XP_017894761.1 5 - 6 251 85

View Full Product Details