Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TET2
Entrez Gene ID
102191595
Gene Name
tet methylcytosine dioxygenase 2
Gene Aliases
-
Chromosome Location
Chr.6: 20216812 - 20352862 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681343.3 XP_005681400.2 5 - 6 4081 62
XM_013964483.2 XP_013819937.2 5 - 6 4081 62
GenBank mRNA KA321926.1 - 6 - 7 3210 62

View Full Product Details