Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRAP
Entrez Gene ID
102190908
Gene Name
GRB2 related adaptor protein
Gene Aliases
-
Chromosome Location
Chr.19: 33858367 - 33879665 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064760.1 XP_017920249.1 3 - 4 606 89
GenBank mRNA JO648481.2 - 3 - 4 360 89

View Full Product Details