Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STARD3NL
Entrez Gene ID
102178696
Gene Name
STARD3 N-terminal like
Gene Aliases
-
Chromosome Location
Chr.4: 70230413 - 70290260 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047198.1 XP_017902687.1 6 - 7 875 75
XM_018047199.1 XP_017902688.1 6 - 7 631 75

View Full Product Details