Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXA9
Entrez Gene ID
102169549
Gene Name
homeobox A9
Gene Aliases
-
Chromosome Location
Chr.4: 51331210 - 51337700 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013963366.2 XP_013818820.2 1 - 2 4011 64

View Full Product Details