Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRSF4
Entrez Gene ID
102176083
Gene Name
serine and arginine rich splicing factor 4
Gene Aliases
-
Chromosome Location
Chr.2: 11572743 - 11598566 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057177.1 XP_017912666.1 4 - 5 897 65
XM_018057187.1 XP_017912676.1 6 - 7 906 65
XM_018057193.1 XP_017912682.1 4 - 5 571 65

View Full Product Details