Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A12
Entrez Gene ID
102170383
Gene Name
solute carrier family 16 member 12
Gene Aliases
-
Chromosome Location
Chr.26: 40149559 - 40250112 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041575.1 XP_017897064.1 8 - 9 2004 74
GenBank mRNA JO425207.1 - 5 - 6 1623 74

View Full Product Details