Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRIQ4
Entrez Gene ID
102181240
Gene Name
leucine rich repeats and IQ motif containing 4
Gene Aliases
-
Chromosome Location
Chr.1: 97064414 - 97081117 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675340.1 XP_005675397.1 3 - 4 1326 63

View Full Product Details