Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBLN7
Entrez Gene ID
102170326
Gene Name
fibulin 7
Gene Aliases
-
Chromosome Location
Chr.11: 124680 - 181748 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054800.1 XP_017910289.1 7 - 8 1253 82
XM_018054801.1 XP_017910290.1 6 - 7 873 82
GenBank mRNA KA334014.1 - 7 - 8 1210 82

View Full Product Details