Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMBX1
Entrez Gene ID
102177681
Gene Name
diencephalon/mesencephalon homeobox 1
Gene Aliases
-
Chromosome Location
Chr.3: 21142661 - 21148198 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046558.1 XP_017902047.1 3 - 4 691 59

View Full Product Details