Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTL3
Entrez Gene ID
102177875
Gene Name
retrotransposon Gag like 3
Gene Aliases
ZCCHC5
Chromosome Location
Chr.X|NW_017189516: 1872642 - 1874360 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700632.2 XP_005700689.2 1 - 2 172 95

View Full Product Details