Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCTE3
Entrez Gene ID
102180227
Gene Name
t-complex-associated-testis-expressed 3
Gene Aliases
-
Chromosome Location
Chr.9: 90988468 - 91000807 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018053496.1 XP_017908985.1 3 - 4 558 104

View Full Product Details