Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABPB2
Entrez Gene ID
102177767
Gene Name
GA binding protein transcription factor subunit beta 2
Gene Aliases
-
Chromosome Location
Chr.3: 100443908 - 100466847 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677630.3 XP_005677687.1 8 - 9 2002 101

View Full Product Details