Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YPEL2
Entrez Gene ID
102189791
Gene Name
yippee like 2
Gene Aliases
-
Chromosome Location
Chr.19: 9933529 - 9998728 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064134.1 XP_017919623.1 2 - 3 472 65
XM_018064135.1 XP_017919624.1 2 - 3 349 65
XM_018064136.1 XP_017919625.1 2 - 3 330 65
XM_018064137.1 XP_017919626.1 2 - 3 384 65
XM_018064139.1 XP_017919628.1 2 - 3 469 65
GenBank mRNA JO425334.1 - 2 - 3 515 65
JO425336.1 - 2 - 3 515 65

View Full Product Details