Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYGN
Entrez Gene ID
102188653
Gene Name
crystallin gamma N
Gene Aliases
-
Chromosome Location
Chr.4: 5881888 - 5887646 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047465.1 XP_017902954.1 2 - 3 272 70

View Full Product Details