Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXHXorf58
Entrez Gene ID
102189020
Gene Name
chromosome X CXorf58 homolog
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 46086882 - 46109875 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013976613.2 XP_013832067.1 7 - 8 987 86

View Full Product Details