Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC108633784
Entrez Gene ID
108633784
Gene Name
gastrin-like
Gene Aliases
-
Chromosome Location
Chr.24: 26860761 - 26861100 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039386.1 XP_017894875.1 1 - 1 312 87

Gene Symbol
GAST
Entrez Gene ID
102179807
Gene Name
gastrin
Gene Aliases
-
Chromosome Location
Chr.19: 41570987 - 41571528 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064071.1 XP_017919560.1 2 - 2 317 87

View Full Product Details