Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Edem2
- Entrez Gene ID
-
34714
- Gene Name
- ER degradation enhancer, mannosidase alpha-like 2
- Gene Aliases
-
CG5682, Dmel\CG5682, Dmel_CG5682, EDEM2, dEDEM2, edem2
- Chromosome Location
-
Chr.2L: 13192201 - 13197281 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032480
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135767.2 |
NP_609611.1 |
6 - 7 |
2461 |
87 |
GenBank mRNA |
BT056246.1 |
- |
6 - 7 |
2458 |
87 |
View Full Product Details