Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CapaR
Entrez Gene ID
40393
Gene Name
Capability receptor
Gene Aliases
CAP2B-R, CG14575, Dm CG14575, Dmel\CG14575, Dmel_CG14575, capaR
Chromosome Location
Chr.3L: 21558974 - 21562181 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037100
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275246.1 NP_001262175.1 1 - 2 390 74
NM_206418.3 NP_996140.1 1 - 2 390 74
GenBank mRNA AF505865.1 - 1 - 2 572 74
AF522193.1 - 1 - 2 209 74

View Full Product Details