Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nmda1
Entrez Gene ID
36419
Gene Name
N-methyl-D-aspartate receptor-associated protein
Gene Aliases
CG3798, DRONMDA, Dmel\CG3798, Dmel_CG3798, NMDA1, NMDARA1, cDNA2, dNMDARA1, dTmbim3
Chromosome Location
Chr.2R: 12877094 - 12880230 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013305
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259359.1 NP_001246288.1 3 - 4 562 68
NM_001259360.2 NP_001246289.1 3 - 4 469 68
NM_001274000.1 NP_001260929.1 1 - 2 427 68
NM_078998.4 NP_523722.1 3 - 4 536 68
NM_165956.3 NP_725237.1 3 - 4 467 68
NM_165957.2 NP_725238.1 3 - 4 566 68
NM_165958.2 NP_725239.1 3 - 4 635 68
NM_165959.3 NP_725240.1 2 - 3 953 68
NM_165960.2 NP_725241.2 3 - 4 577 68
GenBank mRNA AY060346.1 - 2 - 3 890 68
BT001413.1 - 3 - 4 577 68
BT003175.1 - 3 - 4 403 68
BT015290.1 - 3 - 4 388 68

View Full Product Details