Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmat
Entrez Gene ID
3346192
Gene Name
Vesicular monoamine transporter
Gene Aliases
CG33528, CG6119, CG6139, DVMAT, DVMAT-A, DVMAT-B, Dmel\CG33528, Dmel_CG33528, VMAT, Vmt, dVMAT, l(2)SH0459, l(2)SH2 0459, vmat
Chromosome Location
Chr.2R: 13513132 - 13534251 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260964
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014524.2 NP_001014524.1 4 - 5 1205 62
NM_001014525.3 NP_001014525.1 3 - 4 1035 62
NM_001014526.2 NP_001014526.1 4 - 5 1205 62
NM_001274027.2 NP_001260956.1 4 - 5 1021 62
GenBank mRNA BT001408.1 - 3 - 4 1010 62
BT003611.1 - 4 - 5 1188 62
BT011335.1 - 4 - 5 1134 62
BT056262.1 - 4 - 5 1205 62

View Full Product Details