Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Galphao
Entrez Gene ID
36104
Gene Name
G protein alpha o subunit
Gene Aliases
Bkh, CG2204, DG[[o]], DGalpha0, DGalpha[[o]], DROGPAD, Dmel\CG2204, Dmel_CG2204, G(o)alpha47A, G-alpha-47A, G-oalpha, G-oalpha47A, G-oalpha47a, G[[o]], G[[o]]alpha, G[[oalpha]], Galpha47A, Galpha[[047A]], Galpha[[0]], Galpha[[o]], Galpha[[o]]47A, Galphai/o, Galphao47A, Go, Go-alpha, Goa, Goalpha, Goalpha47a, alpha(o), bkh, dGalphao, dgo, dgoalpha, drogpad, goalpha, like GNAO
Chromosome Location
Chr.2R: 10437985 - 10465701 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001122
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078960.5 NP_523684.2 3 - 4 688 77
NM_165772.4 NP_724934.1 3 - 4 570 77
NM_165773.3 NP_724935.1 4 - 5 625 77
NM_176124.3 NP_788304.1 3 - 4 497 77
NM_176125.3 NP_788305.1 3 - 4 552 77
NM_176126.3 NP_788306.1 3 - 4 470 77
NM_176127.3 NP_788307.1 3 - 4 580 77
NM_206079.2 NP_995801.1 2 - 3 700 77
NM_206080.2 NP_995802.1 2 - 3 452 77
GenBank mRNA BT046134.2 - 3 - 4 705 77

View Full Product Details