Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ACXB
- Entrez Gene ID
-
53427
- Gene Name
- Adenylyl cyclase X B
- Gene Aliases
-
AC, AC34A, Ac34A, CG17174, DACXB, Dmel\CG17174, Dmel_CG17174
- Chromosome Location
-
Chr.2L: 12916814 - 12921008 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040509
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138989.3 |
NP_620474.2 |
4 - 5 |
1271 |
81 |
View Full Product Details