Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myo28B1
Entrez Gene ID
53515
Gene Name
Myosin 28B1
Gene Aliases
28B, CG6976, DmVIIB, Dmel\CG6976, Dmel_CG6976, Myo18, Myo28B, VIIb 28B, myo 28B1, myo28B
Chromosome Location
Chr.2L: 7666047 - 7689459 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040299
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144373.4 NP_652630.2 2 - 3 473 70
NM_164758.2 NP_723294.1 2 - 3 328 70
NM_164759.3 NP_723295.2 2 - 3 328 70
NM_164760.4 NP_723296.2 2 - 3 473 70
GenBank mRNA BT133443.1 - 2 - 3 474 70

View Full Product Details