Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Strn-Mlck
Entrez Gene ID
36753
Gene Name
Stretchin-Mlck
Gene Aliases
143736_at, A(225), BEST:GH22543, BcDNA:RH74685, CG15699, CG18255, CG44162, CG8304, CT41348, Dmel\CG44162, Dmel_CG44162, MLCK, MLCK-1, MLCK-2, MLCK-3, Mlc-k, Mlck, Strn-MLCK, Strn-mlck, Strn/Mlck, mlck, strn-mlck
Chromosome Location
Chr.2R: 15946933 - 15989498 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265045
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202026.2 NP_001188955.2 9 - 10 17885 79
NM_001274089.1 NP_001261018.1 9 - 10 17885 79
NM_001274090.1 NP_001261019.1 9 - 10 18253 79
NM_166129.3 NP_725510.2 9 - 10 18253 79

View Full Product Details