Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrj
Entrez Gene ID
36797
Gene Name
CG8448-PA
Gene Aliases
CG8448, Dmel\CG8448, Dmel_CG8448, MRJ, dMRJ, dmrj, mrjA
Chromosome Location
Chr.2R: 16203329 - 16221793 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034091
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_166152.3 NP_725541.1 3 - 4 349 74
NM_166153.3 NP_725542.1 4 - 5 546 74
NM_166154.3 NP_725543.1 3 - 4 563 74
NM_166155.3 NP_725544.1 3 - 4 406 74
GenBank mRNA AY119581.1 - 3 - 4 458 74

View Full Product Details