Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Npc1b
- Entrez Gene ID
-
33072
- Gene Name
- Niemann-Pick type C-1b
- Gene Aliases
-
151, 4, 7, AF2/19, CG12092, Dmel\CG12092, Dmel_CG12092, NPC1b, Q256, R-9-28, R9-28, dncp1b, l(1)19Ec, l(1)2-3, l(1)R-9-28, l151, l152, lN5, lt2-14a, npc1b
- Chromosome Location
-
Chr.X: 20934869 - 20939503 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261675
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134573.3 |
NP_608417.2 |
2 - 3 |
1566 |
154 |
View Full Product Details