Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BBS8
- Entrez Gene ID
-
33217
- Gene Name
- Bardet-Biedl syndrome 8
- Gene Aliases
-
BBS2, CG13691, Dmel\CG13691, Dmel_CG13691
- Chromosome Location
-
Chr.2L: 422873 - 425321 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031255
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134680.3 |
NP_608524.1 |
1 - 2 |
972 |
96 |
View Full Product Details