Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
halo
Entrez Gene ID
33334
Gene Name
CG7428-PB
Gene Aliases
CG7428, Dmel\CG7428, Dmel_CG7428
Chromosome Location
Chr.2L: 1517533 - 1518148 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001174
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144293.2 NP_001137765.1 1 - 1 517 101

View Full Product Details