Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mfr
Entrez Gene ID
39016
Gene Name
misfire
Gene Aliases
CG5747, Dmel\CG5747, Dmel_CG5747, Otoferlin
Chromosome Location
Chr.3L: 8905918 - 8912671 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266757
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144443.2 NP_001137915.1 8 - 9 4039 134
NM_001144444.2 NP_001137916.1 8 - 9 4528 134
NM_001144445.2 NP_001137917.1 7 - 8 3998 134
NM_001144446.2 NP_001137918.1 7 - 8 4527 134
NM_001144447.2 NP_001137919.1 8 - 9 4065 134
NM_001144448.2 NP_001137920.1 7 - 8 4111 134
NM_001144449.2 NP_001137921.1 7 - 8 3975 134

View Full Product Details