Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nwk
Entrez Gene ID
39052
Gene Name
nervous wreck
Gene Aliases
CG43479, CG4641, CG4684, Dmel\CG43479, Dmel_CG43479, NWK, Nwk
Chromosome Location
Chr.3L: 9137860 - 9145693 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263456
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259760.2 NP_001246689.1 4 - 5 903 59
NM_001259761.2 NP_001246690.1 4 - 5 903 59
NM_001259762.2 NP_001246691.1 4 - 5 978 59
NM_001259763.2 NP_001246692.1 4 - 5 903 59
NM_001300064.1 NP_001286993.1 4 - 5 903 59
NM_001300065.1 NP_001286994.1 4 - 5 903 59
NM_001300066.1 NP_001286995.1 4 - 5 903 59
NM_001300067.1 NP_001286996.1 4 - 5 978 59
NM_001300068.1 NP_001286997.1 4 - 5 903 59
NM_140033.3 NP_648290.3 4 - 5 903 59

View Full Product Details