Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
santa-maria
Entrez Gene ID
34024
Gene Name
scavenger receptor acting in neural tissue and majority of rhodopsin is absent
Gene Aliases
BEST:CK01577, CG12789, CK01577, Dmel\CG12789, Dmel_CG12789, Santa Maria, santa maria
Chromosome Location
Chr.2L: 7445355 - 7451244 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025697
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135277.3 NP_609121.2 3 - 4 4568 73
NM_164752.4 NP_723277.3 4 - 5 1307 73

View Full Product Details