Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myo28B1
Entrez Gene ID
53515
Gene Name
Myosin 28B1
Gene Aliases
28B, CG6976, DmVIIB, Dmel\CG6976, Dmel_CG6976, Myo18, Myo28B, VIIb 28B, myo 28B1, myo28B
Chromosome Location
Chr.2L: 7666047 - 7689459 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040299
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_164758.2 NP_723294.1 1 - 2 63 64
NM_164759.3 NP_723295.2 1 - 2 63 64

View Full Product Details