Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNCF
Entrez Gene ID
39496
Gene Name
SoxNeuro Co-Factor
Gene Aliases
BcDNA:RE54375, CG14112, Dmel\CG14112, Dmel_CG14112, HL6, SCNF
Chromosome Location
Chr.3L: 13378583 - 13379190 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036349
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_140378.3 NP_648635.1 1 - 1 470 84
GenBank mRNA AY071463.1 - 1 - 1 468 84

View Full Product Details