Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RhoBTB
Entrez Gene ID
40249
Gene Name
Rho-related BTB domain containing
Gene Aliases
28574635, CG5701, Dmel\CG5701, Dmel_CG5701, RhoGTPase, RhoX
Chromosome Location
Chr.3L: 20376242 - 20382398 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036980
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275178.1 NP_001262107.1 3 - 4 685 62
NM_001275179.1 NP_001262108.1 3 - 4 423 62
NM_140959.2 NP_649216.2 3 - 4 685 62
GenBank mRNA AF217287.1 - 3 - 4 662 62
AY051710.1 - 3 - 4 652 62

View Full Product Details