Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt1
Entrez Gene ID
33473
Gene Name
Synaptotagmin 1
Gene Aliases
CG3139, D. Syt I, DSYT, DSYT1, DSYT2, DSypt, DSypt1, DSyt, DSyt1, Dmel\CG3139, Dmel_CG3139, Droso1, Dsyt2, STY1, SYT, SYT I, SYT1, Syt, Syt 1, Syt I, Syt-1, SytI, dSYT, dSyt1, dsyt1, dsytI, l(2)23AB1, l(2)23Ba, l(2)k05909, syn, syt, syt 1, syt I, syt-1, syt1, sytI
Chromosome Location
Chr.2L: 2772327 - 2799979 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004242
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273036.1 NP_001259965.1 7 - 8 2218 100
NM_001298638.1 NP_001285567.1 10 - 11 1660 100
NM_001298639.1 NP_001285568.1 9 - 10 1657 100
NM_001370000.1 NP_001356906.1 10 - 11 2263 100
NM_078736.3 NP_523460.2 9 - 10 1663 100
NM_164501.2 NP_722838.1 9 - 10 1657 100
NM_164502.2 NP_722839.1 8 - 9 1306 100
NM_205897.2 NP_995619.2 8 - 9 1291 100

View Full Product Details