Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gal
- Entrez Gene ID
-
33839
- Gene Name
- beta galactosidase
- Gene Aliases
-
CG9092, DmelGal, Dmel\CG9092, Dmel_CG9092, beta-GAL, beta-Gal-1, beta-gal, dbeta-Gal, gal, lacZ-1
- Chromosome Location
-
Chr.2L: 6003574 - 6006827 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001089
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135134.3 |
NP_608978.2 |
1 - 2 |
261 |
62 |
GenBank mRNA |
AY119076.1 |
- |
1 - 2 |
225 |
62 |
View Full Product Details