Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
barr
Entrez Gene ID
35287
Gene Name
barren
Gene Aliases
38B.3, Barr, Barren, CAP-H, CG10726, CG14684, Cap-H, CapH, Dmel\CG10726, Dmel_CG10726, dCap-H
Chromosome Location
Chr.2L: 20058163 - 20061370 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014127
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299165.1 NP_001286094.1 2 - 3 788 74
NM_057758.5 NP_477106.1 2 - 3 460 74
NM_165317.3 NP_724240.1 1 - 2 564 74
GenBank mRNA BT011446.1 - 1 - 2 525 74
BT011474.1 - 2 - 3 458 74

View Full Product Details