Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dx
Entrez Gene ID
31589
Gene Name
deltex
Gene Aliases
CG3929, DMDELTEX, Dmel\CG3929, Dmel_CG3929, Dx
Chromosome Location
Chr.X: 6533999 - 6538575 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000524
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078509.3 NP_511064.2 2 - 3 2179 76
GenBank mRNA BT033059.1 - 2 - 3 2144 76

View Full Product Details